missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Calcineurin A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Calcineurin A Polyclonal specifically detects Calcineurin A in Human, Mouse samples. It is validated for Western Blot.Specifications
| Calcineurin A | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway | |
| calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform | |
| PPP3CA | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P63329 | |
| 5530 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title