missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Calcein Blue AM Viability Dye
Description
- Molecular weight: 465.41g/mol
- Peak excitation: 360nm
- Peak emission: 445nm
- Reconstitute in high-quality, freshly opened DMSO
- Once reconstituted, store at −20°C with dessicant and use within a short period of time. Avoid freeze-thawing
- Calcein Blue is optimally excited by the UV laser line (360nm), but can also be excited by the violet laser line (405nm)
- For fluorescent microscopy, detect using the appropriate filter sets
- Calcein Blue AM is itself weakly fluorescent, and an additional wash step may be necessary to reduce background fluorescence
- Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
Specifications
Specifications
| Color | Blue |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Emission | 360/450 |
| Product Line | eBioscience |
| Quantity | 1 mg |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?