missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Calcein AM Viability Dye (UltraPure Grade)

Product Code. 15560597
Change view
Click to view available options
Quantity:
1 mg
20 x 50 μg
Unit Size:
1mg
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15560597 20 x 50 μg Pack of 20
15550597 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15560597 Supplier Invitrogen™ Supplier No. 65085378
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A membrane-permeable live-cell labeling dye. Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein fluorescent dye.

  • Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein fluorescent dye
  • Apoptotic and dead cells with compromised cell membranes do not retain Calcein
  • Calcein is optimally excited at 495 nm and has a peak emission of 515 nm
  • For flow cytometric analysis, it can be excited off the blue laser line (488 nm) and detected using filters for FITC/Alexa Fluor™ 488 (530/30)
  • For multicolor flow cytometry, concentrations in the range of 5 to 20 nM are recommended to ensure that compensation can be performed successfully
  • Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
  • If single-color analysis is desired, improved resolution between live and dead/apoptotic cells may be obtained by using either Calcein Blue AM (cat. 65-0855) or Calcein Violet 450 AM (cat. 65-0854)
  • Calcein AM should be reconstituted in high-quality, freshly opened DMSO
  • Once reconstituted, it should be stored at -20°C with dessicant and used within a short period of time (avoid freeze-thawing)
  • Purity: >95% (determined by HPLC)
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Emission 495/515
Product Line eBioscience
Quantity 20 x 50 μg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.