missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Calcein AM Viability Dye (UltraPure Grade)
Description
- Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein fluorescent dye
- Apoptotic and dead cells with compromised cell membranes do not retain Calcein
- Calcein is optimally excited at 495 nm and has a peak emission of 515 nm
- For flow cytometric analysis, it can be excited off the blue laser line (488 nm) and detected using filters for FITC/Alexa Fluor™ 488 (530/30)
- For multicolor flow cytometry, concentrations in the range of 5 to 20 nM are recommended to ensure that compensation can be performed successfully
- Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
- If single-color analysis is desired, improved resolution between live and dead/apoptotic cells may be obtained by using either Calcein Blue AM (cat. 65-0855) or Calcein Violet 450 AM (cat. 65-0854)
- Calcein AM should be reconstituted in high-quality, freshly opened DMSO
- Once reconstituted, it should be stored at -20°C with dessicant and used within a short period of time (avoid freeze-thawing)
- Purity: >95% (determined by HPLC)
Specifications
Specifications
| Color | Green |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Emission | 495/515 |
| Product Line | eBioscience |
| Quantity | 1 mg |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?