Learn More
Abcam Calcein AM, fluorescent dye for cell viability
Product Code. 32116496 Shop All Abcam Products
Change view
Click to view available options
50 μg
Product Code.
32116496 50 μg
1 options
Product Code. 32116496 Supplier Abcam Supplier No. ab14142050ug
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MW 994.9 Da, Purity >96%. Cell permeant probe used to determine cell viability in most eukaryotic cells. In live cells the nonfluorescent calcein AM is converted to a green-fluorescent calcein after acetoxymethyl ester hydrolysis by intracellular esterases.
MW 994.9 Da, Purity >96%. Cell permeant probe used to determine cell viability in most eukaryotic cells. In live cells the nonfluorescent calcein AM is converted to a green-fluorescent calcein after acetoxymethyl ester hydrolysis by intracellular esterases.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 50 μg