missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Calcein

Product Code. 16408886
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16408886

Brand: MedChemExpress HYD0040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Calcein is a fluorescent dye and self-quenching probe, used as an indicator of lipid vesicle leakage, and also as a complexometric indicator for titration of calcium ions with EDTA, and for fluorometric determination of calcium.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (160.63 mM) ∣ 0.1 M NaOH : 10 mg/mL (16.06 mM; ultrasonic and adjust pH to 11 with NaOH)
Target HBV
Molecular Weight (g/mol) 622.53 g/mol
CAS 1461-15-0
Synonym Fluorexon
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.