missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-4/R-Cadherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£156.00 - £301.00
Specifications
| Antigen | Cadherin-4/R-Cadherin |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18196862
|
Novus Biologicals
NBP2-27372SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129212
|
Novus Biologicals
NBP2-27372 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadherin-4/R-Cadherin Polyclonal specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Mammal samples. It is validated for Western Blot.Specifications
| Cadherin-4/R-Cadherin | |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| CDH4 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen for the antibody is conserved in most mammals. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Mammalia | |
| P55283 | |
| 1002 | |
| Amino acids DYDYLNDWGPRFkkLADMYGGGDD of human Cadherin was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title