missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-4/R-Cadherin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27372
This item is not returnable.
View return policy
Description
Cadherin-4/R-Cadherin Polyclonal specifically detects Cadherin-4/R-Cadherin in Human, Mouse, Mammal samples. It is validated for Western Blot.
Specifications
| Cadherin-4/R-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CAD4, cadherin 4, type 1, preproprotein, cadherin 4, type 1, R-cadherin (retinal), cadherin-4, FLJ22202, FLJ40547, MGC126700, MGC138355, RCAD, R-CAD, R-cadherin, Retinal cadherin | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 1002 | |
| Human, Mouse, Mammalia | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000 | |
| P55283 | |
| CDH4 | |
| Amino acids DYDYLNDWGPRFkkLADMYGGGDD of human Cadherin was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for the antibody is conserved in most mammals. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction