missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-60069
This item is not returnable.
View return policy
Description
Cadherin-22 Polyclonal specifically detects Cadherin-22 in Human samples. It is validated for Western Blot.
Specifications
| Cadherin-22 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CDH22 | |
| Synthetic peptides corresponding to CDH22(cadherin-like 22) The peptide sequence was selected from the N terminal of CDH22. Peptide sequence LIDELTGDIHAMERLDREQKTFYTLRAQARDRATNRLLEPESEFIIKVQD. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Rabbit: 100%; Rat: 100%; Chicken: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| C20orf25, cadherin 22, type 2, cadherin-22, cadherin-like 22, dJ998H6.1, MGC39564, ortholog of rat PB-cadherin, PB-cadherin, Pituitary and brain cadherin | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 64405 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu