missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D1 Antibody (20A), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
£169.00
Specifications
| Antigen | CACNA2D1 |
|---|---|
| Clone | 20A |
| Dilution | Western Blot 1:500, Flow Cytometry 1:100 - 1:200, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
CACNA2D1 Monoclonal specifically detects CACNA2D1 in Human, Mouse, Rat, Porcine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CACNA2D1 | |
| Western Blot 1:500, Flow Cytometry 1:100 - 1:200, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.05% Sodium Azide | |
| CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1 | |
| CACNA2D1 | |
| IgG2a | |
| Unpurified | |
| Detects 1,4-dihydropyridine (DHP) receptor alpha-2 subunit from skeletal and cardiac muscle. Detects a band at 220 kDa under non-reducing and a 143 kDa band under reducing conditions representing DHP from rabbit skeletal muscle membrane preparations. Immunohistochemical staining of DHP in rabbit skeletal muscle with NB120-2864 results in double rows of discrete punctate staining representing pairs of triads on the opposing sides of the Z-lines. |
| 20A | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P13806 | |
| 781 | |
| Purified rabbit dihydropyridine receptor. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title