missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA2D1 Antibody (20A), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB120-2864-0.025ml
This item is not returnable.
View return policy
Description
CACNA2D1 Monoclonal specifically detects CACNA2D1 in Human, Mouse, Rat, Porcine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CACNA2D1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.05% Sodium Azide | |
| CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| 781 | |
| Human | |
| Ascites |
| Western Blot | |
| 20A | |
| Western Blot 1:500, Flow Cytometry 1:100 - 1:200, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation, Immunohistochemistry-Paraffin 1:500, Immunohistochemistry-Frozen 1:500 | |
| P13806 | |
| CACNA2D1 | |
| Purified rabbit dihydropyridine receptor. | |
| 0.025 mL | |
| Primary | |
| Detects 1,4-dihydropyridine (DHP) receptor alpha-2 subunit from skeletal and cardiac muscle. Detects a band at 220 kDa under non-reducing and a 143 kDa band under reducing conditions representing DHP from rabbit skeletal muscle membrane preparations. Immunohistochemical staining of DHP in rabbit skeletal muscle with NB120-2864 results in double rows of discrete punctate staining representing pairs of triads on the opposing sides of the Z-lines. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction