missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1S Mouse anti-Guinea Pig, Human, Mouse, Rabbit, Rat, Clone: 1A, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£159.00
Specifications
| Antigen | CACNA1S |
|---|---|
| Clone | 1A |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen 1:200 |
| Applications | Western Blot |
Description
CACNA1S Monoclonal specifically detects CACNA1S in Human, Mouse, Rat, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| CACNA1S | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Signal Transduction | |
| PBS (pH 7.4) with 0.05% Sodium Azide | |
| 779 | |
| Purified rabbit muscle T-tubule dihydropyridine receptor. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1A | |
| Western Blot 1:500, Flow Cytometry 1 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CACH1, CACN1, CACNL1A3MHS5, calcium channel, L type, alpha 1 polypeptide, isoform 3 (skeletal muscle, Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, calcium channel, voltage-dependent, L type, alpha 1S subunit, Cav1.1, CCHL1A3, dihydropyridine receptor, dihydropyridine-sensitive L-type calcium channel alpha-1 subunit, HOKPP, HOKPP1, hypokalemic periodic paralysis), hypoPP, TTPP1, voltage-dependent L-type calcium channel subunit alpha-1S, Voltage-gated calcium channel subunit alpha Cav1.1 | |
| CACNA1S | |
| IgG1 | |
| Protein A purified | |
| Detects 1,4-dihydropyridine (DHP) receptor alpha-1 subunit in skeletal muscle. The DHP Receptor alpha-1 protein is also known as CACNA1S or Cav1.1 alpha-1 subunit. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title