missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1S Mouse anti-Guinea Pig, Human, Mouse, Rabbit, Rat, Clone: 1A, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-542-50ul
This item is not returnable.
View return policy
Description
CACNA1S Monoclonal specifically detects CACNA1S in Human, Mouse, Rat, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CACNA1S | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen 1:200 | |
| CACH1, CACN1, CACNL1A3MHS5, calcium channel, L type, alpha 1 polypeptide, isoform 3 (skeletal muscle, Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, calcium channel, voltage-dependent, L type, alpha 1S subunit, Cav1.1, CCHL1A3, dihydropyridine receptor, dihydropyridine-sensitive L-type calcium channel alpha-1 subunit, HOKPP, HOKPP1, hypokalemic periodic paralysis), hypoPP, TTPP1, voltage-dependent L-type calcium channel subunit alpha-1S, Voltage-gated calcium channel subunit alpha Cav1.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Detects 1,4-dihydropyridine (DHP) receptor alpha-1 subunit in skeletal muscle. The DHP Receptor alpha-1 protein is also known as CACNA1S or Cav1.1 alpha-1 subunit. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1A | |
| Unconjugated | |
| PBS (pH 7.4) with 0.05% Sodium Azide | |
| CACNA1S | |
| Purified rabbit muscle T-tubule dihydropyridine receptor. | |
| 50 μL | |
| Neuroscience, Signal Transduction | |
| 779 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction