missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1E Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £401.00
Specifications
| Antigen | CACNA1E |
|---|---|
| Dilution | Western Blot 5 - 10 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18360443
|
Bio-Techne
NBP3-11901-25UG |
25 μg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347423
|
Novus Biologicals
NBP3-11901-100UG |
100 μg |
£401.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CACNA1E Polyclonal antibody specifically detects CACNA1E in Human samples. It is validated for Western BlotSpezifikation
| CACNA1E | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Brain calcium channel II, CACH6calcium channel, R type, alpha-1 polypeptide, CACNL1A6calcium channel, voltage-dependent, alpha 1E subunit, calcium channel, voltage-dependent, R type, alpha 1E subunit, Cav2.3, L type, alpha-1 polypeptide, L type, alpha-1 polypeptide, isoform 6, voltage-dependent calcium channel alpha 1E subunit, voltage-dependent R-type calcium channel subunit alpha-1E, voltage-gated calcium channel alpha 1E subunit, Voltage-gated calcium channel subunit alpha Cav2.3 | |
| Synthetic peptide derived from the rat alpha1E calcium channel conjugated to KLH (Uniprot: Q07652) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 5 - 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 777 | |
| IgG | |
| Ammonium sulfate precipitation |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts