missing translation for 'onlineSavingsMsg'
Learn More

CACNA1E Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18360443
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360443 25 μg 25µg
18347423 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18360443 Supplier Bio-Techne Supplier No. NBP31190125UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CACNA1E Polyclonal antibody specifically detects CACNA1E in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA1E
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 5 - 10 ug/ml
Formulation PBS
Gene Alias Brain calcium channel II, CACH6calcium channel, R type, alpha-1 polypeptide, CACNL1A6calcium channel, voltage-dependent, alpha 1E subunit, calcium channel, voltage-dependent, R type, alpha 1E subunit, Cav2.3, L type, alpha-1 polypeptide, L type, alpha-1 polypeptide, isoform 6, voltage-dependent calcium channel alpha 1E subunit, voltage-dependent R-type calcium channel subunit alpha-1E, voltage-gated calcium channel alpha 1E subunit, Voltage-gated calcium channel subunit alpha Cav2.3
Host Species Rabbit
Immunogen Synthetic peptide derived from the rat alpha1E calcium channel conjugated to KLH (Uniprot: Q07652)
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 777
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.