missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CACNA1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30941-100ug

 View more versions of this product

Product Code. 30066813

  • £386.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CACNA1A Polyclonal antibody specifically detects CACNA1A in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CACNA1A
Polyclonal
Western Blot : 1:400, Immunohistochemistry
APCA, BI, brain calcium channel 1, Brain calcium channel I, CACH4, CACN3, CACNL1A4CAV2.1, calcium channel, L type, alpha-1 polypeptide, Calcium channel, L type, alpha-1 polypeptide isoform 4, calcium channel, voltage-dependent, P/Q type, alpha 1A subunit, Cav2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6, voltage-dependent P/Q-type calcium channel subunit alpha-1A, Voltage-gated calcium channel subunit alpha Cav2.1
Synthetic peptide derived from the rat CACNA1A conjugated to KLH
100 μg
Signal Transduction
773
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunohistochemistry, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Antigen Affinity-purified
RUO
Primary
Human, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.