missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CACNA1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | CACNA1A |
|---|---|
| Dilution | Western Blot : 1:400, Immunohistochemistry |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066813
|
Novus Biologicals™
NBP3-30941-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067048
|
Novus Biologicals™
NBP3-30941-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNA1A Polyclonal antibody specifically detects CACNA1A in Human,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| CACNA1A | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| Phosphate buffered saline | |
| 773 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot : 1:400, Immunohistochemistry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| APCA, BI, brain calcium channel 1, Brain calcium channel I, CACH4, CACN3, CACNL1A4CAV2.1, calcium channel, L type, alpha-1 polypeptide, Calcium channel, L type, alpha-1 polypeptide isoform 4, calcium channel, voltage-dependent, P/Q type, alpha 1A subunit, Cav2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6, voltage-dependent P/Q-type calcium channel subunit alpha-1A, Voltage-gated calcium channel subunit alpha Cav2.1 | |
| Synthetic peptide derived from the rat CACNA1A conjugated to KLH | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title