missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £512.00
Specifications
| Antigen | CAC1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656300
|
Novus Biologicals
NBP2-76539-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18360770
|
Novus Biologicals
NBP2-76539 |
100 μL |
£512.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CAC1 Polyclonal specifically detects CAC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CAC1 | |
| Polyclonal | |
| Rabbit | |
| C10orf46, CDK2-associated, cullin domain 1, chromosome 10 open reading frame 46, cullin, FLJ40409, MGC33215 | |
| CACUL1 | |
| IgG | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 143384 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ASSTININTSTSKFLMNVITIEDYKSTYWPKLDGAIDQLLTQSPGDYIPISYEQIYSCVYKCVCQQHSEQMY | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title