missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABP1 Antibody (SR1754), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CABP1 |
|---|---|
| Clone | SR1754 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632675
|
Novus Biologicals
NBP3-22623-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648594
|
Novus Biologicals
NBP3-22623-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CABP1 Monoclonal antibody specifically detects CABP1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| CABP1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9478 | |
| IgG | |
| Affinity purified |
| SR1754 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| CaBP1, Calbrain, calcium binding protein 1, calcium binding protein 5, calcium-binding protein 1, caldendrin, HCALB_BR | |
| A synthesized peptide derived from human CABP1 (Uniprot #: Q9NZU7) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title