missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CABP1 Antibody (SR1754), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22623-50ul
This item is not returnable.
View return policy
Description
CABP1 Monoclonal antibody specifically detects CABP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CABP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9478 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1754 | |
| Western Blot 1:500-1:2000 | |
| CaBP1, Calbrain, calcium binding protein 1, calcium binding protein 5, calcium-binding protein 1, caldendrin, HCALB_BR | |
| A synthesized peptide derived from human CABP1 (Uniprot #: Q9NZU7) | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction