Learn More
Fisherbrand™ Cable for Electrode S7 or S8 Head
Product Code. 11756398 Shop All Fisherbrand Products
Click to view available options
Connector Types:
DIN
Unit Size:
Each
Product Code. 11756398
Only null left
Add to basket
Add to basket
Cable accessory for electrodes with S7 or S8 head is ideal for measuring pH in many lab and scientific environments.
  • Coaxial connector for DIN plug 19262
  • For combined electrodes
  • S7 Cable DIN connector, 3M
  • Knick, WTW, Polymetron, Schott, Sidel
Specifications
Connector Types DIN
For Use With (Equipment) Electrodes with S7 or S8 head
Type Cable accessory
Length (Metric) Cable 3 m