missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Code Corporation™ 6 ft. Straight RS232 Cable
Click to view available options
For Use With (Equipment):
Code Readers
Unit Size:
Each
Description
- Reader termination: 10-pin RJ45
- Host termination: 9-pin, female, D-Sub connector
Specifications
Specifications
| For Use With (Equipment) | Code Readers |
| Type | Cable |
| Alternate Part Number | CRA-C502 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction