missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAB39L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74079
This item is not returnable.
View return policy
Description
CAB39L Polyclonal specifically detects CAB39L in Human samples. It is validated for Western Blot.
Specifications
| CAB39L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Antigen MLAA-34, bA103J18.3, calcium binding protein 39-like, calcium-binding protein 39-like, FLJ12577, MO25-BETA, Mo25-like protein, MO2L, RP11-103J18.3, sarcoma antigen NY-SAR-79, U937-associated antigen | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: . | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9H9S4 | |
| CAB39L | |
| Synthetic peptides corresponding to the N terminal of CAB39L. Immunizing peptide sequence EILCGTNEKEPPTEAVAQLAQELYSSGLLVTLIADLQLIDFEGKKDVTQI. | |
| Affinity purified | |
| RUO | |
| 81617 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction