missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (OV185-1), Novus Biologicals™
Mouse Monoclonal Antibody
£163.00 - £422.00
Specifications
| Antigen | CA125/MUC16 |
|---|---|
| Clone | OV185-1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246484
|
Novus Biologicals
NBP1-22564-0.1ML |
0.1 mL |
£163.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116969
|
Novus Biologicals
NBP1-22564 |
0.5 mL |
£422.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CA125/MUC16 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| MUC16 | |
| IgG1 | |
| Protein A or G purified | |
| This antibody reacts specifically with CA125 ovarian cancer antigen. Several studies have shown that CA125 is a useful tumor marker for ovarian epithelial malignancies. However, CA125 has been described in other neoplasms such as seminal vesicle and anaplastic lymphomas. |
| OV185-1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q8WXI7 | |
| 94025 | |
| Partially purified human mucin fraction from a pool of cancer tissues of patients with epithelial ovarian cancer. | |
| Primary | |
| Store at 4°C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title