missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (OV185-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22564
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 94025 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OV185-1 | |
| Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Q8WXI7 | |
| MUC16 | |
| Partially purified human mucin fraction from a pool of cancer tissues of patients with epithelial ovarian cancer. | |
| 0.5 mL | |
| Primary | |
| This antibody reacts specifically with CA125 ovarian cancer antigen. Several studies have shown that CA125 is a useful tumor marker for ovarian epithelial malignancies. However, CA125 has been described in other neoplasms such as seminal vesicle and anaplastic lymphomas. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction