missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C8A Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£161.00 - £369.00

Specifications

Antigen C8A
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17958942
View Documents
Invitrogen™
PA5145448
20 μL
£161.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17968852
GREENER_CHOICE
View Documents
Invitrogen™
PA5145447
200 μL
£369.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C8A is a component of the complement system and contains three polypeptide subunits: alpha, beta and gamma. C8A, in particular, contains the alpha subunit of C8 and is a constituent of the membrane attack complex (MAC) that plays a key role in the innate and adaptive immune response by forming pores in the plasma membrane of target cells. C8A inserts into the target membrane, but does not form pores by itself. Disease associated with C8A include Type 1 C8 deficiency and immunodeficiency due to C8A deficiency.
Specifications

Specifications

C8A
Western Blot
Unconjugated
Rabbit
Rat
C8a; complement component 8 alpha subunit; complement component 8 subunit alpha; complement component 8, alpha polypeptide; complement component C8 alpha chain
C8A
IgG
Antigen affinity chromatography, Protein A
C8A
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
298288
Recombinant Rat C8a protein, Gln21-His300 (Accession #D3ZWD6).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.