missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C8A Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145447

 View more versions of this product

Product Code. 17968852

  • £369.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

C8A is a component of the complement system and contains three polypeptide subunits: alpha, beta and gamma. C8A, in particular, contains the alpha subunit of C8 and is a constituent of the membrane attack complex (MAC) that plays a key role in the innate and adaptive immune response by forming pores in the plasma membrane of target cells. C8A inserts into the target membrane, but does not form pores by itself. Disease associated with C8A include Type 1 C8 deficiency and immunodeficiency due to C8A deficiency.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C8A
Polyclonal
Unconjugated
C8A
C8A
Recombinant Rat C8a protein, Gln21-His300 (Accession #D3ZWD6).
200 μL
Primary
Rat
Antibody
IgG
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
C8a; complement component 8 alpha subunit; complement component 8 subunit alpha; complement component 8, alpha polypeptide; complement component C8 alpha chain
Rabbit
Antigen affinity chromatography, Protein A
RUO
298288
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.