missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | C5orf4 |
|---|---|
| Concentration | 0.45 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821595
|
Proteintech
22046-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811595
|
Proteintech
22046-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MW of this protein is 25 kDa, and this antibody specifically recognizes the 25 kDa protein.
FAXDC2 gene ontology annotations related to this gene include sterol biosynthetic process.Specifications
| C5orf4 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96IV6 | |
| 10826 | |
| C5orf4 Fusion Protein Ag16960 | |
| Primary | |
| -20°C | |
| FAXDC2 |
| 0.45 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C5orf4, fatty acid hydroxylase domain containing 2, FAXDC2, Uncharacterized protein C5orf4 | |
| FAXDC2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title