missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22046-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 25 kDa, and this antibody specifically recognizes the 25 kDa protein.
FAXDC2 gene ontology annotations related to this gene include sterol biosynthetic process.Specifications
| C5orf4 | |
| Polyclonal | |
| Unconjugated | |
| FAXDC2 | |
| C5orf4, fatty acid hydroxylase domain containing 2, FAXDC2, Uncharacterized protein C5orf4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10826 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96IV6 | |
| FAXDC2 | |
| C5orf4 Fusion Protein Ag16960 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction