missing translation for 'onlineSavingsMsg'
Learn More

Consort C3010/C3030 Multiparameter Dual Channel Analyzers

Product Code. p-7147292
Click to view available options
Product Type:
Multiparameter Analyzer
Total Meter Kit
pH Meter Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11935438

Brand: Consort C3010P

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Two independent channels for all measurements. Consort C3010/C3030 Multiparameter Dual Channel Analyzers are full parameter benchtop meters with unique measurement hardware for each channel, separate temperature inputs, data logging and a USB/RS232 interface for connection to/control by a PC.

  • Multichannel up to 2 measurements can be performed at the same time (2X/sec)
  • Multiparameter for pH, mV, conductivity, resistivity, salinity, TDS, dissolved oxygen, temperature, ion (3030 only), and air pressure
  • No interference between pH/ORP/Ion and conductivity electrodes in the same solution
  • Custom calibration tables for buffer/standard-temperature relations (via device menu or from PC)
  • Wide display shows 2 channels at the same time, including temperature and date/time
  • Stability indicator ensures visualization when the measurement has stabilized
  • Stability algorithm ensures stable readings with ability to detect fast changes
  • Gold plated BNC connectors each accept different electrodes; gold plating prevents corrosion
  • Hold function allows to freeze the display
  • Selectable resolution for more stable readings of mV, pH and DO
  • Range lock for conductivity measurements
  • Built-in help system helps guide users through all steps of calibration and use
  • Capacitive compensation eliminates electrode/cable capacitive component at low conductivities
  • Galvanic isolated USB interface eliminates ground loop effects when connected to a PC
  • GPL report can be shown on the display or sent to the digital port
  • Pre-programmed standards:
    pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 (at 25°C)
    Conductivity: 1413 μS/cm, 12.88 mS/cm, 111.8 mS/cm (at 25°C)
  • Free software and firmware updates downloadable from www.consort.be

Specifications

Outputs USB/RS232
Accuracy (Conductivity) 0.5% f.s. of range
Accuracy (DO) 1% ± 1 digit
Accuracy (mV) 0.1% ± 1 digit
Accuracy (pH) 0.1% ±1 digit
Backlight Option White back light
Calibration GLP
Calibration Points 1 (mV, DO, temperature, air pressure)
Calibration Points (Conductivity) 1 to 3
Calibration Points (pH) 1 to 5
Cell Constant 0.07 to 13 cm-1
Data Management Details Measurement + ° C/date/time
Data Points 12,000
Channels 2 measurement, 2 temperature
Display Type LCD, 240 x 64 pixel
Range (Conductivity) 0 to 2000 mS/cm
Range (mV) ±2000.0 mV
Range (pH) -2.000 to 16.000
Range (Resistivity) 0 to 200 Mohm/cm
Range (Salinity) 0.0 to 70.0
Range (TDS) 0.0 to 100.0 g/L
Resolution (pH) 0.001
Temperature Range (Metric) -5.0°C to +105.0°C
For Use With (Application) pH, mV, conductivity, resistivity, salinity, TDS, DO, air pressure, temperature
Includes pH Kit: C3010 Meter, pH/ATC electrode SP20T, buffers (pH 4 and 7), electrolyte (3 m KCl)
Inputs 2 BNC (measurement), 2 x 2 banana (for Pt1000 for temperature)
Languages English, Dutch, French, German
Model C3010
Pressure Range 600 - 1300 hPa
Product Type pH Meter Kit
Range (DO) 0.00 - 60.00 mg/L
Relative Humidity Range 0 - 95%, non-condensing
Resolution (Conductivity) 0.001 μS/cm
Resolution (DO) 0.01 mg/L
Resolution (mV) 0.1 mV
Resolution (Resistivity) 1 Ω.cm
Resolution (TDS) 0.01 mg/L
Salinity Factor Ref temp: 15° C
Temperature Accuracy 0.1°C
Temperature Compensation −5.0° to +105.0° C (pH, conductivity), 0° to 50° C (DO)
Temperature (Metric) Operating 0°C to 40°C
Temperature (Metric) Reference 20°/25° C (conductivity)
Temperature Resolution 0.1° C
Warranty 36 Months
Dimensions (L x W x H) 26W x 18D x 9H cm
Weight (Metric) 1 kg
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.