missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf55 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-70468
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
C2orf55 Polyclonal specifically detects C2orf55 in Human samples. It is validated for Western Blot.
Specifications
| C2orf55 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| KIAA1211L | |
| Synthetic peptides corresponding to C2ORF55 The peptide sequence was selected from the middle region of C2ORF55. Peptide sequence ITVTRQKRRGTLDQPPNQEDKPGARTLKSEPGKQAKVPERGQEPVKQADF. | |
| Affinity purified | |
| RUO | |
| 343990 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| chromosome 2 open reading frame 55, hypothetical protein LOC343990, KIAA1211L, KIAA1211-like | |
| Rabbit | |
| 102 kDa | |
| 100 μL | |
| Primary | |
| Rat 79%, Canine 86%. | |
| Human, Rat, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction