Learn More
Invitrogen™ C14orf130 Monoclonal Antibody (K49039_3A12), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | C14orf130 |
|---|---|
| Clone | K49039_3A12 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203527
|
Invitrogen™
MA556145 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30205212
|
Invitrogen™
MA555765 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: YPDPEDEIPD EMIQCVVCED WFHGRHLGAI PPESGDFQEM VCQACMKRCS FLWAYAAQLA VTKISTEDDG LVRNIDGIGD QEVIKPENGE HQDSTLKEDV PEQGKDDVRE VKVEQNSEPC AGSSSESDLQ TVFKNESLNA ESKSGCKLQE LKAKQLIKKD TATYWPLNWR SKLCTCQDCM KMYGDLDVLF LTDEYDTVLA YENKGKIAQA TDRSDPLMDT LSSMNRVQQV ELICEYNDLK TELKDYLKRF ADEGTVVKRE DIQQFFEEFQ SKKRRRVDGM
C14orf130 Monoclonal antibody specifically detects C14orf130 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| C14orf130 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 5730410I19Rik; AA589405; AW557761; C14orf130; C21H14ORF130; fb74a06; hypothetical protein LOC322815; LOW QUALITY PROTEIN: putative E3 ubiquitin-protein ligase UBR7; N-recognin-7; Putative E3 ubiquitin-protein ligase UBR7; RGD1359144; RING-type E3 ubiquitin transferase UBR7; ubiquitin protein ligase E3 component n-recognin 7; ubiquitin protein ligase E3 component n-recognin 7 (putative); Ubr7; wu:fb74a06; wu:fi29b06; zgc:55404 | |
| Recombinant human UBR7 (immunogen range is 141-420). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| UBR7 |
| K49039_3A12 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8N806 | |
| 55148 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.