Learn More
Invitrogen™ C14orf130 Monoclonal Antibody (K49039_3A12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555765
Description
Sequence of this protein is as follows: YPDPEDEIPD EMIQCVVCED WFHGRHLGAI PPESGDFQEM VCQACMKRCS FLWAYAAQLA VTKISTEDDG LVRNIDGIGD QEVIKPENGE HQDSTLKEDV PEQGKDDVRE VKVEQNSEPC AGSSSESDLQ TVFKNESLNA ESKSGCKLQE LKAKQLIKKD TATYWPLNWR SKLCTCQDCM KMYGDLDVLF LTDEYDTVLA YENKGKIAQA TDRSDPLMDT LSSMNRVQQV ELICEYNDLK TELKDYLKRF ADEGTVVKRE DIQQFFEEFQ SKKRRRVDGM
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway, recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
| C14orf130 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8N806 | |
| UBR7 | |
| Recombinant human UBR7 (immunogen range is 141-420). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| K49039_3A12 | |
| Unconjugated | |
| UBR7 | |
| 5730410I19Rik; AA589405; AW557761; C14orf130; C21H14ORF130; fb74a06; hypothetical protein LOC322815; LOW QUALITY PROTEIN: putative E3 ubiquitin-protein ligase UBR7; N-recognin-7; Putative E3 ubiquitin-protein ligase UBR7; RGD1359144; RING-type E3 ubiquitin transferase UBR7; ubiquitin protein ligase E3 component n-recognin 7; ubiquitin protein ligase E3 component n-recognin 7 (putative); Ubr7; wu:fb74a06; wu:fi29b06; zgc:55404 | |
| Mouse | |
| Protein A | |
| RUO | |
| 55148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.