Learn More
Palintest™ 8000 Model Photometer
Product Code. 11968527
Click to view available options
Unit Size:
Each
Product Code. 11968527
Brand: Palintest™ PT800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to provide accurate and fast analysis for even the busiest test labs, this Photometer 8000 sets a new standard for water quality testing and ease of operation.
  • High precision instrument with advanced optics technology
  • Rugged design with no moving parts
  • Automatic method set up and wavelength setting
  • Full range of pre-programmed clean and waste water tests
  • Remote software update by email
  • Large, backlit touchscreen
  • Unique adaptive cell holder takes all reagent tubes 12mm to 20mm in diameter
  • Interface translated into five languages
  • On board help with all test instructions
  • Covers the full range of tests for clean and wastewater, it is compatible with all Palintest tablet and liquid photometer tests
    • Accuracy: ±0.005 at 0.3AU
    • Dimensions [w x d x h], mm 290 x 240 x 90
Specifications
Accuracy (pH) Accuracy: ±0.005 at 0.3AU
Description 8000 Model Photometer
Product Type Photometer