missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-myc promoter-binding protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | C-myc promoter-binding protein |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619890
|
Novus Biologicals
NBP2-92291-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628200
|
Novus Biologicals
NBP2-92291-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C-myc promoter-binding protein Polyclonal antibody specifically detects C-myc promoter-binding protein in Human, Mouse samples. It is validated for Western BlotSpecifications
| C-myc promoter-binding protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| c-myc promoter binding protein, C-myc promoter-binding protein, DENN domain-containing protein 4A, DENN/MADD domain containing 4A, IRLBFLJ33949, KIAA0476, MYCPBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1500-1650 of human DENND4A (NP_005839.3). RPGRYFLKSSPSTENMHFPSSISSQTRQSCISTSASGLDTSALSVQGNFDLNSKSKLQENFCTRSIQIPANRSKTAMSKCPIFPMARSISTSGPLDKEDTGRQKLISTGSLPATLQGATDSLGLEWHLPSPDPVTVPYLSPLVVWKELESL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 10260 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title