missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-myc promoter-binding protein Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92291-0.02ml
This item is not returnable.
View return policy
Description
C-myc promoter-binding protein Polyclonal antibody specifically detects C-myc promoter-binding protein in Human, Mouse samples. It is validated for Western Blot
Specifications
| C-myc promoter-binding protein | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| c-myc promoter binding protein, C-myc promoter-binding protein, DENN domain-containing protein 4A, DENN/MADD domain containing 4A, IRLBFLJ33949, KIAA0476, MYCPBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1500-1650 of human DENND4A (NP_005839.3). RPGRYFLKSSPSTENMHFPSSISSQTRQSCISTSASGLDTSALSVQGNFDLNSKSKLQENFCTRSIQIPANRSKTAMSKCPIFPMARSISTSGPLDKEDTGRQKLISTGSLPATLQGATDSLGLEWHLPSPDPVTVPYLSPLVVWKELESL | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10260 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction