missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Mouse, Clone: 9E-10, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB102-100ug
Additional Details : Weight : 0.01000kg
Specifications
c-Myc | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Mouse | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
9E-10 | |
Unconjugated | |
NP_002458 | |
Protein A/G Purified | |
RUO | |
4609 | |
IgG1 κ |