missing translation for 'onlineSavingsMsg'
Learn More

c-Myc Mouse, Clone: 9E-10, enQuireBio™

Mouse Monoclonal Antibody

£99.15 - £173.00

Specifications

Antigen c-Myc
Clone 9E-10
Concentration 0.5mg/mL
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15951488
View Documents Promotion Details
EnquireBio
QAB102-25UG
25 μg
£99.15
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15941488
View Documents Promotion Details
EnquireBio
QAB102-100UG
100 μg
£173.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

c-Myc
0.5mg/mL
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
4609
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
9E-10
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
RUO
NP_002458
IgG1 κ
Protein A/G Purified
Videos
SDS
Documents

Documents

Product Certifications