missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-MAF Monoclonal Antibody (sym0F1), eFluor™ 450, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£149.00 - £386.00
Specifications
| Antigen | c-MAF |
|---|---|
| Clone | sym0F1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16310944
|
eBioscience
48-9855-41 |
25 Tests | |||||||
16320944
![]() |
Invitrogen™
48-9855-42 |
100 Tests |
£386.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The sym0F1 monoclonal antibody reacts with human and mouse c-Maf, a 42 kDa basic leucine zipper transcription factor shown to be involved in the neural, ocular and hematopoietic systems. In hematopoietic cells, it was first shown to be crucial for IL-4 ex
c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet, p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.Specifications
| c-MAF | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| O75444, P54843 | |
| IgG2b κ | |
| 4° C, store in dark, DO NOT FREEZE! | |
| MAF |
| sym0F1 | |
| Flow Cytometry | |
| eFluor 450 | |
| Mouse | |
| Human, Mouse | |
| 17132, 4094 | |
| Primary | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title