missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-MAF Monoclonal Antibody (sym0F1), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: eBioscience 48-9855-41
This item is not returnable.
View return policy
Description
The sym0F1 monoclonal antibody reacts with human and mouse c-Maf, a 42 kDa basic leucine zipper transcription factor shown to be involved in the neural, ocular and hematopoietic systems. In hematopoietic cells, it was first shown to be crucial for IL-4 ex
eFluor 450 is a alternative to Pacific Blue. eFluor 450 emits at 445 nm and is excited with the violet laser line (405 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet , p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.
Specifications
| c-MAF | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
| O75444, P54843 | |
| Maf | |
| Affinity Chromatography | |
| RUO | |
| 17132, 4094 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| sym0F1 | |
| eFluor 450 | |
| MAF | |
| 2810401A20Rik; A230108G15Rik; AW047063; AYGRP; c-MAF; CCA4; CTRCT21; MAF; Maf2 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction