missing translation for 'onlineSavingsMsg'
Learn More

c-Kit/CD117 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen c-Kit/CD117
Concentration 0.23 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898564
View Documents
Proteintech
18696-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888564
View Documents
Proteintech
18696-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KIT may be involved in testicular germ cell tumor (TGCT). A common maligcy in males representing 95% of all testicular neoplasms. TGCTs have various pathologic subtypes including: unclassified intratubular germ cell neoplasia, seminoma (including cases with syncytiotrophoblastic cells), spermatocytic seminoma, embryonal carcinoma, yolk sac tumor, choriocarcinoma, and teratoma. Note=The gene represented in this entry may be involved in disease pathogenesis.
Specifications

Specifications

c-Kit/CD117
Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
P10721
3815
Peptide
Primary
-20°C
KIT
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
C Kit, CD117, KIT, PBT, Proto oncogene c Kit, SCFR, Tyrosine protein kinase Kit
KIT
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.