missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Mouse anti-Human, Mouse, Rat, Clone: 2G9C3, Novus Biologicals™
Mouse Monoclonal Antibody
£157.00
Specifications
| Antigen | c-Fos |
|---|---|
| Clone | 2G9C3 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 2.5-5 ug/ml |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
Description
c-Fos Monoclonal specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| c-Fos | |
| 0.2 mg/mL | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Mouse, Human, Rat | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog | |
| FOS | |
| IgG2a | |
| Protein A or G purified | |
| Reacts with both cellular and viral forms, will detect both the 55 kDa and 62 kDa c-fos and v-fos proteins, but does not react with the 39 kDa jun protein. |
| 2G9C3 | |
| Western Blot 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 2.5-5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| P01100 | |
| 2353 | |
| Synthetic peptide: KVEQLSPEEEEKRRIRRERNKMAAA, corresponding to Internal sequence amino acids 128 - 152 of Human c-Fos. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title