missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Mouse anti-Human, Mouse, Rat, Clone: 2G9C3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1428-0.025MG
This item is not returnable.
View return policy
Description
c-Fos Monoclonal specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| c-Fos | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 2.5-5 ug/ml | |
| P01100 | |
| FOS | |
| Synthetic peptide: KVEQLSPEEEEKRRIRRERNKMAAA, corresponding to Internal sequence amino acids 128 - 152 of Human c-Fos. | |
| 0.025 mg | |
| Apoptosis, Cancer, Hypoxia, Transcription Factors and Regulators, Tumor Suppressors | |
| 2353 | |
| Mouse, Human, Rat | |
| Purified |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 2G9C3 | |
| Unconjugated | |
| 50mM Sodium Phosphate (pH 7.5) and 0.2% Gelatin with 0.1% Sodium Azide | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Reacts with both cellular and viral forms, will detect both the 55 kDa and 62 kDa c-fos and v-fos proteins, but does not react with the 39 kDa jun protein. | |
| Store at 4C. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction