missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-C1 Heavy Chain Receptor Binding Domain Antibody, R&D Systems™

Sheep Polyclonal Antibody
£156.00 - £410.00
Specifications
| Antigen | BoNT-C1 Heavy Chain |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18733698
![]() |
R&D Systems
AF5425-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078381
|
R&D Systems
AF5425 |
100 μg |
£410.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BoNT-C1 Heavy Chain Polyclonal specifically detects BoNT-C1 Heavy Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| BoNT-C1 Heavy Chain | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| C. botulinum | |
| BoNTC1 Heavy Chain, DcD | |
| CbC4_4096 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P18640 | |
| 10523893 | |
| E. coli-derived recombinant BoNT-C1 Heavy Chain Asn866-Glu1291 Accession # P18640 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects C. botulinum BoNT-C1 Heavy Chainin direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant (r) BoNT-D-HCR is observed and 5% cross-reactivity with rBoNT-A-HCR, rBoNT-A-LC, and rBoNT-B-LC is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title