missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-C1 Heavy Chain Receptor Binding Domain Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marke: R&D Systems AF5425
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BoNT-C1 Heavy Chain Polyclonal specifically detects BoNT-C1 Heavy Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.
Spezifikation
| BoNT-C1 Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| P18640 | |
| CbC4_4096 | |
| E. coli-derived recombinant BoNT-C1 Heavy Chain Asn866-Glu1291 Accession # P18640 | |
| 100 μg | |
| Primary | |
| Detects C. botulinum BoNT-C1 Heavy Chainin direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant (r) BoNT-D-HCR is observed and 5% cross-reactivity with rBoNT-A-HCR, rBoNT-A-LC, and rBoNT-B-LC is observed. | |
| C. botulinum | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| BoNTC1 Heavy Chain, DcD | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10523893 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur