missing translation for 'onlineSavingsMsg'
Learn More

Butyrylferrocene, 99%, Thermo Scientific™

Product Code. 11338188
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338188 10 g 10g
11358188 2 g 2g
11348188 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11338188 Supplier Thermo Scientific Alfa Aesar Supplier No. 039404.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-94-9
Molecular Formula C14H16FeO
Molecular Weight (g/mol) 256.13
MDL Number MFCD00143883
InChI Key CVNYMHASFXAXJV-UHFFFAOYNA-N
Synonym Butyroferrocene; Ferrocenyl n-propyl ketone
IUPAC Name Butyrylferrocene
SMILES [Fe].c1cccc1.CCCC(=O)c1cccc1

Specifications

Melting Point 36°C to 40°C
Density 1.254 g/mL
Boiling Point 329°C
Quantity 10 g
Formula Weight 256.13
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Butyrylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.