accessibility menu, dialog, popup

UserName

FeOCH3

CAS RN 1271-94-9

IUPAC Name: (2-butanoylcyclopenta-2,4-dien-1-yl)(cyclopenta-2,4-dien-1-yl)iron
Synonyms: (2-butanoylcyclopenta-2,4-dien-1-yl)(cyclopenta-2,4-dien-1-yl)iron
Molecular Weight (g/mol): 256.13
Molecular Formula: C14H16FeO
InChi Key: CVNYMHASFXAXJV-UHFFFAOYNA-N
SMILES: CCCC(=O)C1=CC=CC1[Fe]C1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results