missing translation for 'onlineSavingsMsg'
Learn More

Butyraldehyde diethylacetal, 97%

Product Code. 10503924 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10503924 25 g Glass bottle 25g
10494635 5 g Glass bottle 5g
10404645 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10503924 Supplier Thermo Scientific Acros Supplier No. 393430250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3658-95-5 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3658-95-5
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00038316
InChI Key UVHXZFGCCJLFMX-UHFFFAOYSA-N
Synonym butane, 1,1-diethoxy, butyraldehyde, diethyl acetal, butyraldehyde diethyl acetal, butylaldehyde diethyl acetal, n-butyraldehyde diethyl acetal, butyraldehyde diethyl acetal, analytical standard
PubChem CID 77225
IUPAC Name 1,1-diethoxybutane
SMILES CCCC(OCC)OCC

Specifications

Color Colorless
Density 0.8300g/mL
Boiling Point 143.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(OCH2CH3)2
Refractive Index 1.3950 to 1.3970 (20°C, 589nm)
Quantity 25 g
Specific Gravity 0.83
Solubility Information Solubility in water: practically insoluble
Formula Weight 146.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Butyraldehyde diethylacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Butyraldehyde diethylacetal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.