CAS RN 3658-95-5
CAS RN 3658-95-5
IUPAC Name:
1,1-diethoxybutane
Synonyms:
1,1-diethoxybutane
Molecular Weight (g/mol):
146.23
Molecular Formula:
C8H18O2
InChi Key:
UVHXZFGCCJLFMX-UHFFFAOYSA-N
SMILES:
CCCC(OCC)OCC
1
–
1
of
1
results
1
Butyraldehyde diethylacetal, 97%
CAS: 3658-95-5 Molecular Formula: C8H18O2 Molecular Weight (g/mol): 146.23 MDL Number: MFCD00038316 InChI Key: UVHXZFGCCJLFMX-UHFFFAOYSA-N Synonym: butane, 1,1-diethoxy,butyraldehyde, diethyl acetal,butyraldehyde diethyl acetal,butylaldehyde diethyl acetal,n-butyraldehyde diethyl acetal,butyraldehyde diethyl acetal, analytical standard PubChem CID: 77225 IUPAC Name: 1,1-diethoxybutane SMILES: CCCC(OCC)OCC