missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Butyl Sepharose™ 4 Fast Flow Chromatography Media
Used for capture and intermediate purification of larger proteins
£148.00 - £612.00
Specifications
Product Type | Butyl Sepharose 4 Fast Flow Media |
---|---|
Matrix | Based on cross-linked 4% agarose |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10317689
|
Cytiva
17-0980-10 |
25 mL |
£148.00
25mL |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
10155917
|
Cytiva
17-0980-01 |
200 mL |
£612.00
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
Description
- Based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
- Hydrophobic interaction chromatography (HIC) media composed of cross-linked 4% agarose
- Aliphatic butyl ligand immobilized to base matrix with ether linkage
- Fast Flow matrix gives the adsorbent high chemical, physical and thermal stabilities
- Dynamic capacity at low salt concentration
- Used with all aqueous buffers commonly used in chromatography
- Autoclavable at 121°C for 20 minutes in water
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
Butyl Sepharose 4 Fast Flow Media | |
Based on cross-linked 4% agarose |